Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azelaic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
123-99-9
C9H16O4
188.223
MFCD00004432
BDJRBEYXGGNYIS-UHFFFAOYSA-N
azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110
2266
CHEBI:48131
nonanedioic acid
C(CCCC(=O)O)CCCC(=O)O
123-99-9
C9H16O4
188.223
MFCD00004432
BDJRBEYXGGNYIS-UHFFFAOYSA-N
azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110
2266
CHEBI:48131
nonanedioic acid
C(CCCC(=O)O)CCCC(=O)O
Assay Percent Range 98%
Linear Formula HOOC(CH2)7COOH
Formula Weight 188.22
Physical Form Slightly Crystalline Powder or Flakes
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Solubility Solubility in water: 2.4g/L water (20°C). Other solubilities: very soluble in alcohol and ether:2.7g/100mL(15°C), soluble in organic solvents, slightly soluble in benzene
Melting Point 98.0°C to 103.0°C
Color White to Yellow
Boiling Point 237.0°C (20.0hPa)
Quantity 25g
Flash Point 215°C
Chemical Name or Material Azelaic acid, 98%
Show More Show Less