Azer Scientific Formagel™ Tissue Fixation and Transport Gel

Solution to leaking formalin problems for improved histology studies

Manufacturer: AZER SCIENTIFIC INC PFG40

 View more versions of this product

Catalog No. 22-026-356

  Check Availability
Add to Cart

    Specifications

    Closure Type Screw-on
    For Use With (Application) Routine, special, IHC staining procedures
    Includes 10% Neutral Buffered Formalin
    Material Polypropylene
    Color Clear,Clear
    Product Type Formagel™ Prefilled Container
    Quantity 20mL,20mL
    Type Formagel™ Prefilled Container

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit