Learn More
Azer Scientific Formalin Absorbent Shipping and Grossing Pads
Absorb and neutralize formalin and glutaraldehyde preventing hazardous vapours
Manufacturer:  AZER SCIENTIFIC INC ES86671
 View more versions of this product
Catalog No. 22-026-236
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Formalin Absorbent Shipping Pad
To absorb and neutralize formalin and glutaraldehyde preventing hazardous vapors
200 Case