Azer Scientific Histology Tissue Control Slides
Designed for use in monitoring stain performance in the pathology laboratory
Manufacturer: AZER SCIENTIFIC INC ESCS171
 View more versions of this product
Catalog No. 22-026-726
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Designed for use in monitoring stain performance in the pathology laboratory
    Includes 10 unstained and 1 stained unifrost slide
    Type CMV Immunohistochemistry Control Slide
    Quantity 11 Pack
  • Description & Specifications
Related Products
Product Content Feedback