Learn More
Newcomer Supply AZF (Acetic Zinc Formalin) Fixative, 1 gallon. Excellent replacement for Mercury based fixatives
SDP
Supplier:  Newcomer Supply 1009B
Encompass
Use: Excellent replacement for Mercury based fixative. Working Solution. MERCURY FREE! Newcomer Supply AZF (Acetic Zinc Formalin) Fixative is a ready-to-use fixative recommended for bone marrow clots and cores, lymph nodes, endoscopic biopsies and immunohistochemical (IHC) studies. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC9753295
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.