Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AZI2/NAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 150421AP150UL
Description
AZI2, or NAP1, contributes to the activation of NFKB (Fujita et al., 2003 ).Specifications
AZI2/NAP1 | |
Polyclonal | |
Unconjugated | |
AZI2 | |
5 azacytidine induced 2, AZ2, AZI2, Nak associated protein 1, NAP1, TILP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64343 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H6S1 | |
AZI2 | |
AZI2/NAP1 Fusion Protein Ag6892 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction