Learn More
Molecular Probes™ Azido, PEO (4 propionic Acid, Succinimidyl Ester (3-(Azidotetra(Ethyleneoxy))Propionic Acid, Succinimidyl Ester)) Non-distribution product as customer accommodation.
Azido, PEO (4 propionic Acid, Succinimidyl Ester (3-(Azidotetra(Ethyleneoxy))Propionic Acid, Succinimidyl Ester))
Manufacturer:  Molecular Probes™ A10280
Catalog No. A10280
Add to cart
Description
Description
Conjugates prepared with the amine-reactive alkyne, succinimidyl ester can be detected with an azide-containing molecule in a click chemistry reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.
Specifications
Specifications
Azido, PEO (4 propionic Acid, Succinimidyl Ester (3-(Azidotetra(Ethyleneoxy))Propionic Acid, Succinimidyl Ester)), 1mg, Colorless, Solid, 388.38 Molecular Weight, C15H24N4O8 Molecular Formula, Azide Dye Type, Used For Labeling Chemistry
Colorless
Azide
≤–20°C
388.38
Solid
Cell analysis, labeling chemistry, labeling amines, amine-reactive fluorophores, click chemistries, cell proliferation with click chemistry
Azido, PEO (4 propionic Acid, Succinimidyl Ester (3-(Azidotetra(Ethyleneoxy))Propionic Acid, Succinimidyl Ester))
1mg
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.