Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azilsartan, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
147403-03-0
C25H20N4O5
456.458
KGSXMPPBFPAXLY-UHFFFAOYSA-N
azilsartan,unii-f9nux55p23,azilsartan tak-536,chembl57242,2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzimidazole-7-carboxylic acid,1h-benzimidazole-7-carboxylic acid, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy,1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy-1h-benzimidazole-7-carboxylic acid,2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzo d imidazole-7-carboxylic acid,2-ethoxy-1-2'-4,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl benzimidazole-7-carboxylic acid,2-ethoxy-3-4-2-5-oxo-2h-1,2,4-oxadiazol-3-yl phenyl phenyl methyl benzimidazole-4-carboxylic acid
9825285
CHEBI:68850
2-ethoxy-3-[[4-[2-(5-oxo-2H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylic acid
CCOC1=NC2=CC=CC(=C2N1CC3=CC=C(C=C3)C4=CC=CC=C4C5=NC(=O)ON5)C(=O)O
This item is not returnable. View return policy
147403-03-0
C25H20N4O5
456.458
KGSXMPPBFPAXLY-UHFFFAOYSA-N
azilsartan,unii-f9nux55p23,azilsartan tak-536,chembl57242,2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzimidazole-7-carboxylic acid,1h-benzimidazole-7-carboxylic acid, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy,1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy-1h-benzimidazole-7-carboxylic acid,2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzo d imidazole-7-carboxylic acid,2-ethoxy-1-2'-4,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl benzimidazole-7-carboxylic acid,2-ethoxy-3-4-2-5-oxo-2h-1,2,4-oxadiazol-3-yl phenyl phenyl methyl benzimidazole-4-carboxylic acid
9825285
CHEBI:68850
2-ethoxy-3-[[4-[2-(5-oxo-2H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylic acid
CCOC1=NC2=CC=CC(=C2N1CC3=CC=C(C=C3)C4=CC=CC=C4C5=NC(=O)ON5)C(=O)O

Potent AT1 receptor inverse agonist; antihypertensive

Potent angiotensin II type 1 (AT1) receptor inverse agonist (IC50 = 2.6 nM at the human AT1 receptor). Inhibits angiotensin II-induced IP1 accumulation in COS-7 cells; decreases maximal contraction of rabbit aortic strips in a concentration-dependent manner (pD'2 = 9.9). Antihypertensive; prevents vascular cell proliferation and expression of PAI-1.
Quantity 50mg
Formula Weight 456.45
Percent Purity >98%
Chemical Name or Material Azilsartan

Recommended Storage : Store at Room Temperature