Learn More
LGC Standards USA Azithromycin for peak identification, 117772-70-0, MFCD01862248, 4mg  DFS Item
Manufacturer:  LGC Standards USA EPY0000637
Molecular Formula C38H72N2O12.2H2O, Molecular Weight 785.018
Catalog No. 50-715-990