Learn More
Dynalon™ Azlon™ Carboys with Stopcock, Heavy-Duty Polypropylene with Graduations
Squat form heavy duty carboy with molded handle
$101.00
Specifications
Certifications/Compliance FDA Compliant
Closure Material Polypropylene
Autoclavable Yes
Graduations Yes
Material Polypropylene
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
S37233 View Documents Dynalon™
5056342
10 L 90mm Each for $101.00
Add to cart
S37232 View Documents Dynalon™
5056341/EMD
5 L 45mm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
S37234 View Documents Dynalon™
5056343/EMD
20 L 90mm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Squat form heavy-duty carboy with stopcock is molded of natural polypropylene and supplied with a screw cap and molded-in carry handle.
  • Range of high-performance autoclavable carboys in a choice of three sizes
  • Autoclavable polypropylene, heavy-duty construction
  • FDA-compliant resin
  • Stylish rounded octagonal shape for increased rigidity when full
  • Offset neck for easier filling and pouring
  • Integral, comfortable blow molded handles for ease of carrying are end-sealed to prevent contamination
  • Heavy-duty closures are compatible with other leading brands
  • Carboys feature a durable and leak-proof heavy-duty spigot with boss fitting for extra strength
  • Spigot features removable tube fitting for added convenience
  • Molded graduations allow easy viewing of contents 
Specifications
Specifications
FDA Compliant
Yes
Polypropylene
Wide
Offset
Polypropylene
Yes
Closure, Handles, Spigot
135°C
Octagonal