Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Azlon™ Solvent Venting Wash Bottles

Manufacturer:  Dynalon™ 5064650002

 View more versions of this product

Catalog No. 03-405-14

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Translucent low density polyethylene wash bottles feature unique two-way valve that stops vapor pressure build-up and eliminates the hazard of volatile solvent leaks. Wide neck provides for easier and safer filling. Plus, for quick identification, cadmium-free polypropylene dispensing closures are color coded. And, for optimum control in dispensing, the jet is precision molded. Each bottle features a clear, indelible label describing the contents, international hazard codes and instructional diagrams for safe usage. Unlabeled bottles with write-on label areas and complete set of adhesive hazard labels for custom use are also available. Supplied in convenient shelf packs of five.

Order Info

Bottles are available in 250 or 500mL capacities with five common solvent labels.

Compliance

All bottles have SPI recycling code stamped on base.

Specifications

Specifications

16 oz., 500mL
16 oz.
Polypropylene
Ethanol
LDPE
72.89mm
Orange
500mL
SPI recycling code stamped on base
Translucent
9 in.
2.87 in.
No
228.6mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit