Learn More
Azo Violet, Spectrum™
Click to view available options
Quantity:
5 g
25 g
100 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
74-39-5
C12H9N3O4
259.22
BIFFKKLAAIGZBV-UHFFFAOYSA-N
3-hydroxy-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O
74-39-5
C12H9N3O4
259.22
BIFFKKLAAIGZBV-UHFFFAOYSA-N
3-hydroxy-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O
AZ115, 74-39-5, C12H9N3O4
Spectrum™ Azo Violet , also known as azoviolet or Magneson, is used as a dye and pH indicator. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g