Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azobenzene 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
500g
103-33-3
C12H10N2
182.226
MFCD00003022
DMLAVOWQYNRWNQ-UHFFFAOYSA-N
azobenzene,diazene, diphenyl,diazobenzene,e-diphenyldiazene,azobisbenzene,azofume,1,2-diphenyldiazene,azodibenzeneazofume,diphenyldiimide,azobenzide
2272
CHEBI:190358
diphenyldiazene
C1=CC=C(C=C1)N=NC2=CC=CC=C2
103-33-3
C12H10N2
182.226
MFCD00003022
DMLAVOWQYNRWNQ-UHFFFAOYSA-N
azobenzene,diazene, diphenyl,diazobenzene,e-diphenyldiazene,azobisbenzene,azofume,1,2-diphenyldiazene,azodibenzeneazofume,diphenyldiimide,azobenzide
2272
CHEBI:190358
diphenyldiazene
C1=CC=C(C=C1)N=NC2=CC=CC=C2
Quantity 25g
Boiling Point 293°C
Melting Point 68°C
Color Undesignated
UN Number 3077
Formula Weight 182.23
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material Azobenzene

EINECSNumber : (3)-0361

RTECSNumber : CN1400000

TSCA : Yes