Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azodicarbonamide, FCC, 98.6-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
123-77-3
C2H4N4O2
116.08
XOZUGNYVDXMRKW-UHFFFAOYSA-N
(carbamoylimino)urea
NC(=O)N=NC(N)=O
123-77-3
C2H4N4O2
116.08
XOZUGNYVDXMRKW-UHFFFAOYSA-N
(carbamoylimino)urea
NC(=O)N=NC(N)=O

A1386, 123-77-3, C2H4N4O2

Spectrum™ Azodicarbonamide, FCC is used as a flour bleaching agent and a dough conditioner. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 2.5 kg
Loss on Drying 0.005
Ignition Residue 0.0015
CAS Max % 1
UN Number UN3242
Percent Purity 98.6 to 100.5%