Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azoic Diazo Component 24 (Base) 97.0+%, TCI America™

Click to view available options
5G
6268-05-9
C15H16N2O3
272.30
MFCD00008369
DDRCIGNRLHTTIW-UHFFFAOYSA-N
N-(4-Amino-2,5-dimethoxyphenyl)benzamide, 4-Benzamido-2,5-dimethoxyaniline, Fast Blue RR Base
80442
N-(4-amino-2,5-dimethoxyphenyl)benzamide
COC1=CC(=C(C=C1N)OC)NC(=O)C2=CC=CC=C2
6268-05-9
C15H16N2O3
272.30
MFCD00008369
DDRCIGNRLHTTIW-UHFFFAOYSA-N
N-(4-Amino-2,5-dimethoxyphenyl)benzamide, 4-Benzamido-2,5-dimethoxyaniline, Fast Blue RR Base
80442
N-(4-amino-2,5-dimethoxyphenyl)benzamide
COC1=CC(=C(C=C1N)OC)NC(=O)C2=CC=CC=C2
Melting Point 170°C
Quantity 5g
Color Gray-Red
Formula Weight 272.30
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (HPLC)
Chemical Name or Material Azoic Diazo Component 24 (Base)

EINECSNumber : (5)-2314

TSCA : Yes