Azomethine-H hydrate, Alfa Aesar™

Manufacturer: Alfa Aesar 1681004

Quantity 2g

 View more versions of this product

Catalog No. AA1681004

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32266-60-7
    Molecular Formula C17H13NO8S2·xH2O
    Formula Weight 423.40 (Anhydrous)
    MDL Number MFCD00066536
    Chemical Name or Material Azomethine-H hydrate

    Specifications

    Formula Weight 423.40 (Anhydrous)
    Quantity 2g
    Physical Form Crystalline
    Molecular Formula C17H13NO8S2·xH2O
    MDL Number MFCD00066536
    CAS 32266-60-7
    Chemical Name or Material Azomethine-H hydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit