Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azoxybenzene 98.0+%, TCI America™

Click to view available options
Quantity:
25g
495-48-7
C12H10N2O
198.225
MFCD00019925
GAUZCKBSTZFWCT-UHFFFAOYSA-N
azoxybenzene,azoxybenzide,azoxybenzol,fenazox,fentoxan,azobenzene, oxide,diphenyldiazene 1-oxide,benzene, azoxydi,diazene, diphenyl-, 1-oxide,ordinary azoxybenzene
10316
CHEBI:51865
oxido-phenyl-phenyliminoazanium
C1=CC=C(C=C1)N=[N+](C2=CC=CC=C2)[O-]
495-48-7
C12H10N2O
198.225
MFCD00019925
GAUZCKBSTZFWCT-UHFFFAOYSA-N
azoxybenzene,azoxybenzide,azoxybenzol,fenazox,fentoxan,azobenzene, oxide,diphenyldiazene 1-oxide,benzene, azoxydi,diazene, diphenyl-, 1-oxide,ordinary azoxybenzene
10316
CHEBI:51865
oxido-phenyl-phenyliminoazanium
C1=CC=C(C=C1)N=[N+](C2=CC=CC=C2)[O-]
Quantity 25g
Melting Point 36°C
Color Undesignated
Boiling Point 130°C
Formula Weight 198.23
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material Azoxybenzene

EINECSNumber : (3)-0362

RTECSNumber : CO4025000

TSCA : Yes

Recommended Storage : Refrigerator