Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Azure I

Click to view available options
Quantity:
5g
25g
100g
531-55-5
C15H16ClN3S
305.824
MFCD00011935
DNDJEIWCTMMZBX-UHFFFAOYSA-N
azure b,azure i,methylene azure b,3-dimethylamino-7-methylamino phenothiazin-5-ium chloride,trimethylthionine chloride,azur i,methyleneazure,methylene azure van,unii-7c8bel8wqv,modr kresylova bb czech
68275
dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride
CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]
531-55-5
C15H16ClN3S
305.824
MFCD00011935
DNDJEIWCTMMZBX-UHFFFAOYSA-N
azure b,azure i,methylene azure b,3-dimethylamino-7-methylamino phenothiazin-5-ium chloride,trimethylthionine chloride,azur i,methyleneazure,methylene azure van,unii-7c8bel8wqv,modr kresylova bb czech
68275
dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride
CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]
Applications
Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie′s modified Nocht′s method for paraffin sections.

Solubility
Soluble in 1 mg/ml water. Sparingly soluble in alcohol.

Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.
Melting Point ~205°C (decomposition)
Odor Odorless
Quantity 100g
Beilstein 3922630
Merck Index 14,6058
Solubility Information Soluble in 1mg/ml water. Sparingly soluble in alcohol.
Formula Weight 305.83
Chemical Name or Material Azure I

GHS H Statement
H341
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 208-511-2

RTECSNumber : SO5550000

TSCA : No

Recommended Storage : Ambient temperatures