Learn More
Moore Medical B Braun™ Ultrasite™ IV Sets
Specifications
Quantity Each
Includes Roller clamp, Ultrasite™ Y-site 3 in. above distal end, Safeline™ Y-site 8 in. above distal end and spinlock connection
Length (English) 84 in.
View More Specs
Products 1
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
19-898-484
View Documents
Moore Medical
72102
15 Drops/mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Specifications
Specifications
Each
Roller clamp, Ultrasite™ Y-site 3 in. above distal end, Safeline™ Y-site 8 in. above distal end and spinlock connection
84 in.