Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappa Goat anti-Mouse, Biotin, Polyclonal, Southern Biotech™
Goat Polyclonal Antibody
Supplier: Southern Biotechnology Associates 105008
Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
Kappa | |
Polyclonal | |
Biotin | |
Goat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG |
ELISA, Dot Blot, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.5mg/mL | |
Supplied in PBS/NaN3 | |
Mouse κ light chains | |
1.0 mg | |
Secondary | |
Store at 2-8°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction