Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B-HT 920, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Dopamine D2 receptor agonist, α2-adrenoceptor agonist and 5-HT3 receptor antagonist. Displays antiParkinsonian activity.
Specifications
Quantity | 50 mg |
Formula Weight | 282.23 |
Percent Purity | >98% |
Chemical Name or Material | B-HT 920 |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature