Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ B I09
IRE1 endonuclease inhibitor; cell permeable
$327.12 - $1365.32
Specifications
Color | Beige |
---|---|
Product Type | Inhibitor |
CAS | 1607803-67-7 |
Molecular Formula | C16H17NO5 |
Formula Weight | 303.31g/mol |
Description
IRE-1 RNase inhibitor (IC50 = 1.23 μM). Inhibits IRE-1/XBP1 pathway. Cell permeable. Inhibits growth of human chronic lymphocytic leukemia (CLL) cells in vitro and promotes CLL regression in a mouse model.Specifications
Beige | |
1607803-67-7 | |
303.31g/mol | |
Store at -20°C |
Inhibitor | |
C16H17NO5 | |
Soluble to 50mM in DMSO | |
Inhibits Rnase |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title