Learn More
Cayman ChemicalSupplier Diversity Partner β-Nicotyrine, 487-19-4, 25 mg DFS Item
Manufacturer:  Cayman ChemicalSupplier Diversity Partner 1633325
An alkaloid metabolite of nicotine as well as a major product of its pyrolysis; binds comparably to CYP2A6 and CYP2A13 (Kis = 7.5 and 5.6 µM, respectively), but appears to more effectively inactivate CYP2A6
Catalog No. NC1468739