Learn More
Molecular Probes™ B-Phycoerythrin Non-distribution product as customer accommodation.
B-Phycoerythrin
Manufacturer:  Molecular Probes™ P800
Catalog No. P800
Add to cart
Description
Description
Phycobiliprotein bioconjugates including antibody and other protein conjugates can be made with the exceptionally bright phycobiliprotein, B-phycoerythrin using the Protein-Protein Crosslinking Kit (P6305).
Specifications
Specifications
Store in refrigerator (2–8°C) and protect from light.
Wet Ice
4 mg⁄ml
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.