Learn More
Thermo Scientific™ B-pure Deionization Systems: Cartridges
Accessory cartridges for deionization systems.
Supplier: Thermo Scientific™ D0803/EMD
Description
- Demineralizer Mult Cartridge for use with B-Pure Deionization System
- Resin type: Mixed bed
- High purity
Specifications
1725 grains as NaCl, 875 | |
B-Pure™ deionization system and analytical lab testing | |
Demineralizer Mult Cartridge |
Mixed Bed | |
High |