Learn More
B-Raf Antibody (OTI5A9), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270260UV
Catalog No. NBP270260UV
Add to Cart
Description
Description
B-Raf Monoclonal specifically detects B-Raf in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
B-Raf
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
BRAF
Full length recombinant protein of human B-Raf (NP_004324) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat, Monkey
IgG1
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI5A9
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109
Mouse
Affinity Purified
Apoptosis, Phospho Specific
673.0
Store at 4°C in the dark.
Recommendations
Recommendations