Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-Raf Mouse anti-Human, Monkey, Alexa Fluor™ 647, Clone: OTI5A9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270260AF647

Catalog No. N270260A647


Add to cart

Description

Description

B-Raf Monoclonal antibody specifically detects B-Raf in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

B-Raf
OTI5A9
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
BRAF
Full length recombinant protein of human B-Raf (NP_004324) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Phospho Specific
Primary
673.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit