Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B-Raf Mouse anti-Human, Monkey, DyLight 755, Clone: OTI5A9, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270260IR
Description
B-Raf Monoclonal antibody specifically detects B-Raf in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence.Specifications
B-Raf | |
OTI5A9 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Affinity Purified, Conjugated | |
BRAF | |
Full length recombinant protein of human B-Raf (NP_004324) produced in HEK293T cells. | |
Immunogen affinity purified | |
Apoptosis, Phospho Specific | |
Primary | |
673.0 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human, Monkey |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title