Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B2, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Promotes inclusion formation in cellular models of Huntington's disease and Parkinson's disease. Prevents mutant huntingtin-mediated proteasome dysfunction and reduces α-synuclein-mediated toxicity.
Specifications
Quantity | 10 mg |
Formula Weight | 396.83 |
Percent Purity | >98% |
Chemical Name or Material | B2 |
Safety and Handling
Recommended Storage : Store at +4°C