Learn More
beta-2 Microglobulin Mouse anti-Human, FITC, Clone: B2M-01, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15737
Catalog No. 50-113-7086
Add to cart
Description
Description
Beta-2-microglobulin (B2M) is a 12-14 kDa protein that is a subunit of the major histocompatibility complex (MHC) class I. Human beta 2 Microglobulin associated with HLA Class I antigens are expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells, but is absent on erythrocytes. Beta-2 microglobulin is also found in free form in the plasma. The amino acid sequence of Beta-2-microglobulin is homologous to the constant regions of immunoglobulin heavy and light chains, with a predomitly beta pleated sheet structure. The protein is ubiquitously expressed in the cytoplasm, and is involved in antigen processing and presentation. Beta-2-microglobulin also plays a role in TCR signaling, and downstream signaling in naive CD8+ cells. Beta 2-microglobulin is encoded by the B2M gene located on chromosome 15. A mutation in the Beta-2-microglobulin gene has been shown to result in hypercatabolic hypoproteinemia. Levels of beta 2 microglobulin can be used to assess renal function in kidney transplant recipients. Beta-2-microglobulin that protein can be used as a prognostic marker for HIV infection.
Specifications
Specifications
beta-2 Microglobulin
Monoclonal
1 mg/mL
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P61769
B2-microglobulin, B2M, CDABP0092, HDCMA22P, beta chain MHC class I molecules, MHC Class 1 beta
Mouse
IgG2a
100 μg
4° C
Primary
567
Flow Cytometry
B2M-01
FITC
B2M
Liquid
B2M
human beta2-microglobulin.
Purified
RUO
Antibody
Monoclonal
Human