B30 Series Heavy-Duty Print Ribbon

“Drop & Click” print ribbons for quick and easy supply changes

Manufacturer: Brady Corporation B30R10000GR8

 View more versions of this product

Catalog No. 19-100-048

  Check Availability
Add to Cart

    Specifications

    Format Cartridge
    Length (English) 200 ft roll
    Print Technology BBP® 31 Printer; BBP® 33 Printer; BBP® 35 Printer; BBP® 37 Printer
    Color Green/Red
    Color Capability 2-Color Ribbon
    Style Ribbon in Cartridge
    Width (English) 4.33 in. ribbon

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit