Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ B3GALT5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563006
Description
Immunogen sequence: YSLNPFKEQS FVYKKDGNFL KLPDTDCRQT PPFLVLLVTS SHKQLAERMA IRQTW Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 68%.
B3GALT5 catalyzes the transfer of Gal to GlcNAc-based acceptors with a preference for the core3 O-linked glycan GlcNAc(beta1,3)GalNAc structure. Can use glycolipid LC3Cer as an efficient acceptor. (Uniprot).
Specifications
B3GALT5 | |
Polyclonal | |
Unconjugated | |
B3GALT5 | |
1190002B21Rik; AU045265; B3GALT5; b3Gal-T5; b3Galt-V; B3GalTx; B3gt5; B3T5; Beta-1,3-galactosyltransferase 5; beta-1,3-GalTase 5; beta-3-galactosyltransferase 5; beta3GalT5; beta3Gal-T5; beta-3-Gx-T5; GLCT5; homolog of C. elegans Bt toxin resistance gene bre-5; SSEA-3 synthase; Stage-specific embryonic antigen 3 synthase; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 5; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 5; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 5; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10317 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2C3 | |
B3GALT5 | |
Recombinant protein corresponding to Human B3GALT5. Recombinant protein control fragment (Product #RP-101480). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction