Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ B3GALTL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529055
Description
Recommended positive controls: 293T, Raji. Predicted reactivity: Mouse (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a beta-1, 3-glucosyltransferase that transfers glucose to O-linked fucosylglycans on thrombospondin type-1 repeats (TSRs) of several proteins. The encoded protein is a type II membrane protein. Defects in this gene are a cause of Peters-plus syndrome (PPS).
Specifications
B3GALTL | |
Polyclonal | |
Unconjugated | |
B3GLCT | |
B3galtl; B3GLCT; B3Glc-T; B3GTL; beta 1,3-galactosyltransferase-like; beta 3-glucosyltransferase; beta 3-glycosyltransferase-like; beta-1,3-glucosyltransferase; Beta3Glc-T; beta-3-glucosyltransferase; beta-3-glycosyltransferase-like; Gal-T; Gm1057; UDP-GAL:beta-GlcNAc beta-1,3-galactosyltransferase-like | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
145173 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q6Y288 | |
B3GLCT | |
Recombinant protein fragment corresponding to a region within amino acids 140 and 380 of Human B3GALTL. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction