Learn More
B3GAT1 Mouse anti-Human, Clone: NK-1, Abnova™
Mouse monoclonal antibody raised against native human B3GAT1.
Supplier: Abnova Corporation MAB13281
Description
The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized. [provided by RefSeq]
Specifications
B3GAT1 | |
Monoclonal | |
Unconjugated | |
Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
CD57/GLCATP/GlcAT-P/GlcUAT-P/HNK-1/HNK1/LEU7/NK-1 | |
Mouse | |
PEG precipitation | |
RUO | |
27087 | |
Store at 4°C. | |
IgM κ |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
NK-1 | |
Mouse monoclonal antibody raised against native human B3GAT1. | |
In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
B3GAT1 | |
Human peripheral blood mononuclear cells. | |
100 μg | |
Primary | |
Human | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.