Learn More
B3GAT1 Mouse anti-Human, Clone: SPM129, Abnova™
Mouse monoclonal antibody raised against native human B3GAT1.
Manufacturer:  Abnova Corporation MAB13031
Catalog No. 89-933-616
Add to cart
Description
Description
The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized. [provided by RefSeq]
Specifications
Specifications
B3GAT1
SPM129
Mouse monoclonal antibody raised against native human B3GAT1.
In 10mM PBS.
CD57/GLCATP/GlcAT-P/GlcUAT-P/HNK-1/HNK1/LEU7/NK-1
Mouse
IgM, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
B3GAT1
Human peripheral blood mononuclear cells.
Polyethylene Glycol (PEG) precipitation
RUO
Primary
27087