Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B3GNT2 Mouse anti-Human, Clone: OTI3A9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526101

Catalog No. PIMA526101


Add to cart

Description

Description

This gene encodes a member of the beta-1,3-N-acetylglucosaminyltransferase family. This enzyme is a type II transmembrane protein. It prefers the substrate of lacto-N-neotetraose, and is involved in the biosynthesis of poly-N-acetyllactosamine chains.
Specifications

Specifications

B3GNT2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NY97
B3GNT2; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2 ; B3GN-T1; B3GN-T2; B3GNT; B3GNT-2; B3GNT1; BETA3GNT; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1; beta-1,3-N-acetylglucosaminyltransferase bGnT-1; beta-1,3-N-a
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10678
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3A9
Unconjugated
B3GNT2
Liquid
B3GNT2
Full length human recombit protein of B3GNT2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit