Learn More
Invitrogen™ B3GNT5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589820
Description
Immunogen sequence: DNHIVSHMKS YSYRYLINSY DFVNDTLSLK HTSAGPRYQY LINHKEKCQA QDVLLLLFVK TAPENYDRRS GIRRTWGNEN YVRSQLNANI KTLFALGTPN PLEGEELQRK LAWEDQRYND IIQQDFVDSF YNLTLKLLMQ FSWANTYCPH AKFLMTADDD IFIHMPNLIE YLQSLEQIGV QDFWIGRVHR GAPPIRDKSS KYYVSYEMYQ WPAYPDYTAG AAYVISGDVA AKVYEASQTL NSSLYIDDVF MGLCANKIGI VPQDHVFFSG EGKTPYHPCI YEKMMTSHGH LEDLQDLWKN ATDPKVKTIS KGFFGQIYCR LMKIILLCKI SYVDTYPCRA AFI; Positive Samples: HT-29, A-549; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.
B3GNT5 is a member of the beta-1,3-N-acetylglucosaminyltransferase family. This enzyme is a type II membrane protein. It exhibits strong activity to transfer GlcNAc to glycolipid substrates and is identified as the most likely candidate for lactotriaosylceramide synthase. This enzyme is essential for the expression of Lewis X epitopes on glycolipids.
Specifications
B3GNT5 | |
Polyclonal | |
Unconjugated | |
B3GNT5 | |
B3gnt5; B3GN-T5; beta 1,3 N-acetyglucosaminyltransferase Lc3 synthase; beta-1,3-Gn-T5; beta-1,3-N-acetylglucosaminyltransferase 5; beta-1,3-N-acetylglucosaminyltransferase bGnT-5; beta-1,3-N-acetylglucosaminyltransferase-5; beta3GnT5; beta3Gn-T5; BGnT-5; lactosylceramide 1,3-N-acetyl-beta-D-glucosaminyltransferase; Lactotriaosylceramide synthase; lc(3)Cer synthase; lc3 synthase; UDP-GlcNAc:beta-Gal beta-1,3-N-acetylglucosaminyltransferase 5; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84002 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.03 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9BYG0 | |
B3GNT5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 36-378 of human B3GNT5 (NP_1144361). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.