Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B4GALNT3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548615

Catalog No. PA548615

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Transfers N-acetylgalactosamine (GalNAc) from UDP-GalNAc to N-acetylglucosamine-beta-benzyl with a beta-1,4-linkage to form N,N ft.-diacetyllactosediamine, GalNAc-beta-1,4-GlcNAc structures in N-linked glycans and probably O-linked glycans. Mediates the N,N ft.- diacetyllactosediamine formation on gastric mucosa.
Specifications

Specifications

B4GALNT3
Polyclonal
PBS with 0.09% sodium azide
Q6L9W6
Beta-1, 4-N-acetylgalactosaminyltransferase 3, B4GalNAcT3, Beta4GalNAc-T3, Beta4GalNAcT3, Beta-1, 4-N-acetylgalactosaminyltransferase III, N-acetyl-beta-glucosaminyl-glycoprotein 4-beta-N-acetylgalactosaminyltransferase 2, NGalNAc-T2, B4GALNT3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
283358
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
B4GALNT3
Liquid
B4GALNT3
KLH conjugated synthetic peptide between 555-584 amino acids from the Central region of human B4GALNT3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit