Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B4GALT3 Antibody (OTI1G9), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271386AF700
Description
B4GALT3 Monoclonal specifically detects B4GALT3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
B4GALT3 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
B4GALT3 | |
Recombinant protein fragment corresponding to amino acids 100-393 of human B4GALT3 (NP_003770) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot | |
OTI1G9 | |
Western Blot | |
b4Gal-T3, beta-1,4-galactosyltransferase 3, Beta-1,4-GalTase 3, Beta4Gal-T3, beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 3, EC 2.4.1.-, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 3, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 3, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 3 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
8703.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction