Learn More
B4GALT4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152001AP
Catalog No. 50-173-0060
Add to cart
Description
Description
This gene is one of seven beta-1,4-galactosyltransferase (beta4GalT) genes. They encode type II membrane-bound glycoproteins that appear to have exclusive specificity for the donor substrate UDP-galactose; all transfer galactose in a beta1,4 linkage to similar acceptor sugars: GlcNAc, Glc, and Xyl. Each beta4GalT has a distinct function in the biosynthesis of different glycoconjugates and saccharide structures. As type II membrane proteins, they have an N-terminal hydrophobic signal sequence that directs the protein to the Golgi apparatus and which then remains uncleaved to function as a transmembrane anchor. By sequence similarity, the beta4GalTs form four groups: beta4GalT1 and beta4GalT2, beta4GalT3 and beta4GalT4, beta4GalT5 and beta4GalT6, and beta4GalT7. The enzyme encoded by this gene appears to mainly play a role in glycolipid biosynthesis. Two alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
B4GALT4
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60513, Q66HH1, Q9JJ04
B4Gal T4, B4GALT4, Beta 1,4 GalTase 4, beta4Gal T4
Rabbit
IgG
150 μL
-20°C
Primary
303923, 56375, 8702
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
B4GALT4
Liquid
B4galt4
B4GALT4 Fusion Protein Ag7348
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat