Learn More
B7-1/CD80 Hamster anti-Mouse, Alexa Fluor™ 750, Clone: AP-MAB0713, Novus Biologicals™
Hamster Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212178AF750
Catalog No. NP212178A70
Add to cart
Description
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
B7-1/CD80
AP-MAB0713
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Hamster
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CD80
Mouse CD80
0.1 mL
Primary
941