Learn More
B7-1/CD80 Hamster anti-Mouse, DyLight 488, Clone: AP-MAB0713, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212178G0.1ML
Catalog No. NB212178G01
Add to cart
Description
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
B7-1/CD80
AP-MAB0713
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CD80
Mouse CD80
>95% pure by SDS-PAGE
RUO
Store at 4C in the dark.
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Hamster
IgG
0.1 ml
Adaptive Immunity, Cytokine Research, Immunology, Signal Transduction
Primary
941