Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-1/CD80, Human anti-Human, Clone: IDEC-114 (Galiximab), Novus Biologicals™

Human Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen B7-1/CD80
Clone IDEC-114 (Galiximab)
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Blocking Assay
Host Species Human
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB016037
View Documents
Novus Biologicals
NBP2810990.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB016038
View Documents
Novus Biologicals
NBP2810990.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human Samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Blocking Assay.
Specifications

Specifications

B7-1/CD80
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Blocking Assay
Human
Primary
IDEC-114 (Galiximab)
Human
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit