Learn More
B7-1/CD80 Mouse anti-Human, Alexa Fluor™ 350, Clone: 62N3G8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225255AF350
Catalog No. NP222A30
Add to cart
Description
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
B7-1/CD80
62N3G8
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD80
Amino acids 102-223 of human CD80 were used as the immunogen for the antibody.
0.1 mL
Primary
941