Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-1/CD80 Mouse anti-Human, Alexa Fluor™ 350, Clone: C80/1608, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254307AF350

Catalog No. NP24307A30


Add to cart

Description

Description

B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human samples. It is validated for Flow Cytometry,ELISA,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

B7-1/CD80
C80/1608
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD80
Recombinant human full-length CD80 protein
0.1 mL
Primary
941
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit