Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-1/CD80 Mouse anti-Human, Mouse, Alexa Fluor 594, Clone: 62N3G8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP225255AF594

Catalog No. NP222A94


Add to cart

Description

Description

B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

B7-1/CD80
62N3G8
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
CD80
Amino acids 102-223 of human CD80 were used as the immunogen for the antibody.
Protein G purified
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction
Primary
941.0
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit