Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-1/CD80 Mouse anti-Human, Clone: C80/1608- Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

$410.59

Specifications

Antigen B7-1/CD80
Clone C80/1608
Host Species Mouse
Isotype IgG1 Kappa
Gene Alias Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254307100
View Documents
Novus Biologicals
NBP254307100UG
100 ug
Each for $410.59
Add to cart
 
NB254307200
View Documents
Novus Biologicals
NBP254307200UG
200 ug This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human samples. It is validated for ELISA, CyTOF-ready.
Specifications

Specifications

B7-1/CD80
Mouse
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF
Unconjugated
Store at -20 to -80C. Avoid freeze-thaw cycles.
Monoclonal
1 mg/mL
C80/1608
IgG1 Kappa
Human
RUO
Purified
Primary
PBS with No Preservative
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit