Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-2/CD86 Rat anti-Human, Mouse, Alexa Fluor™ 594, Clone: GL1, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP230148AF594

Catalog No. NP230148A94


Add to cart

Description

Description

B7-2/CD86 Monoclonal antibody specifically detects B7-2/CD86 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

B7-2/CD86
GL1
Flow Cytometry, Knockout Validated
Conjugated, Purified
CD86
LPS-activated mouse B cells
Protein G purified
Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Dendritic Cell Markers, Diabetes Research, Immunology, Myeloid derived Suppressor Cell
Primary
942.0
Flow Cytometry, Immunoassay
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Activation B7-2 antigen, B70, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Rat
IgG2a Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit