Learn More
B7-2/CD86 Rat anti-Mouse, Alexa Fluor 405, Clone: GL1, R&D Systems™
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB741V100UG
Catalog No. FAB741V100
Add to cart
This item is not returnable. View return policy
Description
Description
B7-2/CD86 Monoclonal antibody specifically detects B7-2/CD86 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
B7-2/CD86
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Rat
IgG2a
RUO
Primary
942
Mouse
Flow Cytometry
GL1
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD86
LPS-activated mouse B cells
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse B7-2/CD86 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse B7-1, recombinant human (rh) B7-1 or rhB7-2 is observed.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)