Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H2/ICOSLG, Mouse anti-Human, Alexa Fluor 350, Clone: ICOSL/3260, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279932AF350

Catalog No. NB012847


Add to cart

Description

Description

B7-H2/ICOSLG Monoclonal antibody specifically detects B7-H2/ICOSLG in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

B7-H2/ICOSLG
ICOSL/3260
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
ICOSLG
Recombinant fragment of human B7-H2/ICOSLG protein (around aa 23-149) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 350
50mM Sodium Borate with 0.05% Sodium Azide
B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand
Mouse
IgG2b Kappa
0.1 mL
Cell Cycle and Replication
Primary
This monoclonal antibody recognizes a protein of 36kDa, identified as B7-H2/ICOSLG.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit